Assay Details
| Gene Symbol: Lage3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875009.3 | XP_004875066.1 | 2 - 3 | 575 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lage3| Gene Symbol: Lage3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875009.3 | XP_004875066.1 | 2 - 3 | 575 | 75 | |
Set Membership: |
|