Assay Details
| Gene Symbol: Abhd17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838135.3 | XP_004838192.2 | 4 - 5 | 1436 | 94 | |
| XM_021252714.1 | XP_021108373.1 | 5 - 6 | 1106 | 94 | ||
| XM_021252715.1 | XP_021108374.1 | 5 - 6 | 1508 | 94 | ||
| XM_021252716.1 | XP_021108375.1 | 5 - 6 | 1038 | 94 | ||
More Information
Set Membership: |
|

Abhd17b
Genomic Map