Assay Details
| Gene Symbol: Tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848175.2 | XP_004848232.1 | 1 - 2 | 212 | 96 | |
| XM_021261180.1 | XP_021116839.1 | 1 - 2 | 212 | 96 | ||
| XM_021261181.1 | XP_021116840.1 | 1 - 2 | 212 | 96 | ||
More Information
Set Membership: |
|

Tmem38b
Genomic Map