Assay Details
| Gene Symbol: Map7d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862333.3 | XP_004862390.2 | 16 - 17 | 2586 | 65 | |
| XM_021242522.1 | XP_021098181.1 | 16 - 17 | 2579 | 65 | ||
More Information
Set Membership: |
|

Map7d3
Genomic Map