Assay Details
Gene Symbol: Slc9a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871554.2 | XP_004871611.1 | 10 - 11 | 1468 | 60 | |
XM_013068064.2 | XP_012923518.1 | 10 - 11 | 1467 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
