Assay Details
| Gene Symbol: Trabd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072083.2 | XP_012927537.2 | 2 - 3 | 248 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trabd2a| Gene Symbol: Trabd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072083.2 | XP_012927537.2 | 2 - 3 | 248 | 70 | |
Set Membership: |
|