Assay Details
| Gene Symbol: Atoh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847128.2 | XP_004847185.1 | 1 | 2523 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004847128.2 transcripts.
Set Membership: |
|

Atoh7
Genomic Map