Assay Details
| Gene Symbol: Wnt5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864328.3 | XP_004864385.2 | 4 - 5 | 1758 | 59 | |
| XM_021244058.1 | XP_021099717.1 | 4 - 5 | 905 | 59 | ||
More Information
Set Membership: |
|

Wnt5a
Genomic Map