Assay Details
| Gene Symbol: Ccdc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867530.3 | XP_004867587.1 | 2 - 3 | 658 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc59| Gene Symbol: Ccdc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867530.3 | XP_004867587.1 | 2 - 3 | 658 | 83 | |
Set Membership: |
|