Assay Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856533.2 | XP_004856590.1 | 3 - 4 | 552 | 59 | |
| XM_004856534.2 | XP_004856591.1 | 3 - 4 | 552 | 59 | ||
| XM_004856536.2 | XP_004856593.1 | 2 - 3 | 528 | 59 | ||
| XM_021237794.1 | XP_021093453.1 | 3 - 4 | 577 | 59 | ||
| XM_021237795.1 | XP_021093454.1 | 4 - 5 | 660 | 59 | ||
| XM_021237796.1 | XP_021093455.1 | 2 - 3 | 367 | 59 | ||
More Information
Set Membership: |
|

Layn
Genomic Map