Assay Details
| Gene Symbol: Tmem95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857403.2 | XP_004857460.1 | 5 - 6 | 606 | 80 | |
| XM_021238344.1 | XP_021094003.1 | 4 - 5 | 525 | 80 | ||
More Information
Set Membership: |
|

Tmem95
Genomic Map