Assay Details
| Gene Symbol: Slc22a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021264659.1 | XP_021120318.1 | 9 - 10 | 2671 | 59 | |
| XM_021264660.1 | XP_021120319.1 | 8 - 9 | 2626 | 59 | ||
| XM_021264661.1 | XP_021120320.1 | 8 - 9 | 2504 | 59 | ||
| XM_021264662.1 | XP_021120321.1 | 7 - 8 | 1779 | 59 | ||
| XM_021264663.1 | XP_021120322.1 | 8 - 9 | 1218 | 59 | ||
More Information
Set Membership: |
|

Slc22a12
Genomic Map