Assay Details
| Gene Symbol: Fam109b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845491.3 | XP_004845548.1 | 2 - 3 | 174 | 86 | |
| XM_013072528.2 | XP_012927982.1 | 2 - 3 | 342 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004845491.3, XM_013072528.2 transcripts.
Set Membership: |
|

Fam109b
Genomic Map