Assay Details
| Gene Symbol: Ccdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867108.2 | XP_004867165.1 | 1 | 1821 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc8| Gene Symbol: Ccdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867108.2 | XP_004867165.1 | 1 | 1821 | 104 | |
Set Membership: |
|