Assay Details
| Gene Symbol: Serpinb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858456.2 | XP_004858513.1 | 2 - 3 | 237 | 62 | |
| XM_021239287.1 | XP_021094946.1 | 2 - 3 | 332 | 62 | ||
| XM_021239288.1 | XP_021094947.1 | 2 - 3 | 224 | 62 | ||
More Information
Set Membership: |
|

Serpinb7
Genomic Map