Assay Details
| Gene Symbol: Shisa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845501.2 | XP_004845558.1 | 2 - 3 | 666 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shisa8| Gene Symbol: Shisa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845501.2 | XP_004845558.1 | 2 - 3 | 666 | 90 | |