Assay Details
| Gene Symbol: Tmem41b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851399.3 | XP_004851456.1 | 1 - 2 | 235 | 64 | |
| XM_004851400.2 | XP_004851457.1 | 2 - 3 | 213 | 64 | ||
| XM_021263779.1 | XP_021119438.1 | 2 - 3 | 222 | 64 | ||
More Information
Set Membership: |
|

Tmem41b
Genomic Map