Assay Details
| Gene Symbol: B3gat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862679.3 | XP_004862736.2 | 4 - 5 | 1261 | 66 | |
| XM_004862680.3 | XP_004862737.2 | 4 - 5 | 1271 | 66 | ||
| XM_013078834.2 | XP_012934288.1 | 4 - 5 | 889 | 66 | ||
| XM_021242761.1 | XP_021098420.1 | 4 - 5 | 1994 | 66 | ||
More Information
Set Membership: |
|

B3gat1
Genomic Map