Assay Details
| Gene Symbol: Znf652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859649.2 | XP_004859706.1 | 3 - 4 | 1161 | 63 | |
| XM_004859650.3 | XP_004859707.1 | 4 - 5 | 1611 | 63 | ||
| XM_021239894.1 | XP_021095553.1 | 4 - 5 | 1431 | 63 | ||
More Information
Set Membership: |
|

Znf652
Genomic Map