Assay Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866802.3 | XP_004866859.1 | 1 - 2 | 339 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866802.3 transcripts.
Set Membership: |
|

Lrrc4b
Genomic Map