Assay Details
| Gene Symbol: Lrrc4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866802.3 | XP_004866859.1 | 2 - 3 | 662 | 71 | |
| XM_004866803.3 | XP_004866860.1 | 2 - 3 | 612 | 71 | ||
| XM_004866804.3 | XP_004866861.1 | 2 - 3 | 1180 | 71 | ||
| XM_004866805.2 | XP_004866862.1 | 2 - 3 | 532 | 71 | ||
| XM_004866806.3 | XP_004866863.1 | 2 - 3 | 521 | 71 | ||
More Information
Set Membership: |
|

Lrrc4b
Genomic Map