Assay Details
| Gene Symbol: Mterf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258365.1 | XP_021114024.1 | 1 - 2 | 1313 | 113 | |
| XM_021258366.1 | XP_021114025.1 | 1 - 2 | 841 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021258365.1, XM_021258366.1 transcripts.
Set Membership: |
|

Mterf2
Genomic Map