Assay Details
| Gene Symbol: Mterf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021258365.1 | XP_021114024.1 | 2 - 3 | 1730 | 76 | |
| XM_021258366.1 | XP_021114025.1 | 2 - 3 | 1258 | 76 | ||
More Information
Set Membership: |
|

Mterf2
Genomic Map