Assay Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861173.2 | XP_004861230.1 | 3 - 4 | 438 | 69 | |
| XM_021241412.1 | XP_021097071.1 | 2 - 3 | 263 | 69 | ||
| XM_021241413.1 | XP_021097072.1 | 3 - 4 | 413 | 69 | ||
| XM_021241414.1 | XP_021097073.1 | 3 - 4 | 500 | 69 | ||
More Information
Set Membership: |
|

Tspan31
Genomic Map