Assay Details
| Gene Symbol: Slx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864754.2 | XP_004864811.1 | 14 - 15 | 6050 | 60 | |
| XM_013065639.2 | XP_012921093.1 | 15 - 16 | 5736 | 60 | ||
| XM_021244408.1 | XP_021100067.1 | 16 - 17 | 5672 | 60 | ||
| XM_021244409.1 | XP_021100068.1 | 15 - 16 | 5510 | 60 | ||
| XM_021244410.1 | XP_021100069.1 | 15 - 16 | 5566 | 60 | ||
| XM_021244411.1 | XP_021100070.1 | 15 - 16 | 5563 | 60 | ||
| XM_021244412.1 | XP_021100071.1 | 14 - 15 | 4791 | 60 | ||
| XM_021244413.1 | XP_021100072.1 | 9 - 10 | 3796 | 60 | ||
More Information
Set Membership: |
|

Slx4
Genomic Map