Assay Details
| Gene Symbol: LOC101723532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859752.3 | XP_004859809.2 | 2 - 3 | 306 | 68 | |
| XM_013077662.2 | XP_012933116.1 | 2 - 3 | 321 | 68 | ||
| XM_013077663.2 | XP_012933117.1 | 2 - 3 | 694 | 68 | ||
| XM_013077665.2 | XP_012933119.1 | 2 - 3 | 154 | 68 | ||
| XM_021240128.1 | XP_021095787.1 | 3 - 4 | 1170 | 68 | ||
| XM_021240129.1 | XP_021095788.1 | 2 - 3 | 153 | 68 | ||
More Information
Set Membership: |
|

LOC101723532
Genomic Map