Assay Details
| Gene Symbol: Naglu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859833.2 | XP_004859890.1 | 5 - 6 | 1082 | 56 | |
| XM_021240069.1 | XP_021095728.1 | 4 - 5 | 937 | 56 | ||
More Information
Set Membership: |
|

Naglu
Genomic Map