Assay Details
| Gene Symbol: Gatad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862369.3 | XP_004862426.1 | 3 - 4 | 772 | 89 | |
| XM_013078733.2 | XP_012934187.1 | 3 - 4 | 917 | 89 | ||
| XM_021242590.1 | XP_021098249.1 | 2 - 3 | 646 | 89 | ||
More Information
Set Membership: |
|

Gatad1
Genomic Map