Assay Details
| Gene Symbol: Cdc42ep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866689.3 | XP_004866746.2 | 1 - 2 | 219 | 84 | |
| XM_021246162.1 | XP_021101821.1 | 1 - 2 | 219 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866689.3, XM_021246162.1 transcripts.
Set Membership: |
|

Cdc42ep5
Genomic Map