Assay Details
| Gene Symbol: Cdc42ep5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866689.3 | XP_004866746.2 | 2 - 3 | 350 | 53 | |
| XM_004866694.3 | XP_004866751.1 | 2 - 3 | 878 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866694.3 transcripts.
Set Membership: |
|

Cdc42ep5
Genomic Map