Assay Details
| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862170.2 | XP_004862227.1 | 3 - 4 | 658 | 57 | |
| XM_013078598.2 | XP_012934052.1 | 4 - 5 | 712 | 57 | ||
| XM_021242395.1 | XP_021098054.1 | 4 - 5 | 712 | 57 | ||
| XM_021242396.1 | XP_021098055.1 | 4 - 5 | 735 | 57 | ||
| XM_021242397.1 | XP_021098056.1 | 2 - 3 | 1458 | 57 | ||
More Information
Set Membership: |
|

Etnk2
Genomic Map