Assay Details
| Gene Symbol: Znf300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075742.2 | XP_012931196.1 | 1 - 2 | 156 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013075742.2 transcripts.
Set Membership: |
|

Znf300
Genomic Map