Assay Details
| Gene Symbol: Cdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242548.1 | XP_021098207.1 | 1 - 2 | 84 | 95 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdr1| Gene Symbol: Cdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021242548.1 | XP_021098207.1 | 1 - 2 | 84 | 95 | |