Assay Details
| Gene Symbol: Slc22a31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256557.1 | XP_021112216.1 | 2 - 3 | 449 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a31| Gene Symbol: Slc22a31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256557.1 | XP_021112216.1 | 2 - 3 | 449 | 72 | |
Set Membership: |
|