Assay Details
Gene Symbol: Muc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834675.3 | XP_004834732.1 | 2 - 3 | 2292 | 78 | |
XM_013071868.2 | XP_012927322.1 | 4 - 5 | 2257 | 78 | ||
XM_021254866.1 | XP_021110525.1 | 4 - 5 | 2306 | 78 | ||
XM_021254868.1 | XP_021110527.1 | 4 - 5 | 2168 | 78 | ||
XM_021254872.1 | XP_021110531.1 | 4 - 5 | 2168 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
