Assay Details
| Gene Symbol: Pdgfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845583.3 | XP_004845640.1 | 2 - 3 | 232 | 65 | |
| XM_004845584.3 | XP_004845641.1 | 2 - 3 | 164 | 65 | ||
| XM_021258983.1 | XP_021114642.1 | 2 - 3 | 232 | 65 | ||
| XM_021258984.1 | XP_021114643.1 | 2 - 3 | 4118 | 65 | ||
More Information
Set Membership: |
|

Pdgfb
Genomic Map