Assay Details
| Gene Symbol: Hsd17b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842786.3 | XP_004842843.1 | 4 - 5 | 992 | 77 | |
| XM_004842787.3 | XP_004842844.1 | 3 - 4 | 809 | 77 | ||
More Information
Set Membership: |
|

Hsd17b2
Genomic Map