Assay Details
| Gene Symbol: Smim14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870291.2 | XP_004870348.2 | 2 - 3 | 136 | 64 | |
| XM_004870292.3 | XP_004870349.1 | 2 - 3 | 347 | 64 | ||
More Information
Set Membership: |
|

Smim14
Genomic Map