Assay Details
| Gene Symbol: Znf185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067909.2 | XP_012923363.1 | 20 - 21 | 2084 | 66 | |
| XM_013067910.2 | XP_012923364.1 | 19 - 20 | 2046 | 66 | ||
| XM_013067911.2 | XP_012923365.1 | 19 - 20 | 1990 | 66 | ||
| XM_013067912.2 | XP_012923366.1 | 6 - 7 | 920 | 66 | ||
| XM_021249157.1 | XP_021104816.1 | 20 - 21 | 2088 | 66 | ||
More Information
Set Membership: |
|

Znf185
Genomic Map