Assay Details
Gene Symbol: Fam107b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013078531.2 | XP_012933985.1 | 4 - 5 | 913 | 63 | |
XM_013078532.2 | XP_012933986.1 | 4 - 5 | 912 | 63 | ||
XM_013078533.2 | XP_012933987.1 | 4 - 5 | 913 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
