Assay Details
| Gene Symbol: Tex15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855457.2 | XP_004855514.2 | 4 - 5 | 699 | 126 | |
| XM_021236851.1 | XP_021092510.1 | 6 - 7 | 941 | 126 | ||
| XM_021236852.1 | XP_021092511.1 | 7 - 8 | 1063 | 126 | ||
| XM_021236853.1 | XP_021092512.1 | 7 - 8 | 999 | 126 | ||
| XM_021236854.1 | XP_021092513.1 | 6 - 7 | 859 | 126 | ||
| XM_021236855.1 | XP_021092514.1 | 5 - 6 | 975 | 126 | ||
| XM_021236856.1 | XP_021092515.1 | 5 - 6 | 865 | 126 | ||
| XM_021236857.1 | XP_021092516.1 | 5 - 6 | 775 | 126 | ||
| XM_021236858.1 | XP_021092517.1 | 6 - 7 | 897 | 126 | ||
| XM_021236859.1 | XP_021092518.1 | 6 - 7 | 833 | 126 | ||
| XM_021236860.1 | XP_021092519.1 | 5 - 6 | 693 | 126 | ||
| XM_021236861.1 | XP_021092520.1 | 4 - 5 | 809 | 126 | ||
More Information
Set Membership: |
|

Tex15
Genomic Map