Assay Details
| Gene Symbol: Tex15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855457.2 | XP_004855514.2 | 7 - 8 | 8132 | 79 | |
| XM_021236851.1 | XP_021092510.1 | 9 - 10 | 8374 | 79 | ||
| XM_021236852.1 | XP_021092511.1 | 10 - 11 | 8496 | 79 | ||
| XM_021236853.1 | XP_021092512.1 | 10 - 11 | 8432 | 79 | ||
| XM_021236854.1 | XP_021092513.1 | 9 - 10 | 8292 | 79 | ||
| XM_021236855.1 | XP_021092514.1 | 8 - 9 | 8408 | 79 | ||
| XM_021236856.1 | XP_021092515.1 | 8 - 9 | 8298 | 79 | ||
| XM_021236857.1 | XP_021092516.1 | 8 - 9 | 8208 | 79 | ||
| XM_021236858.1 | XP_021092517.1 | 9 - 10 | 8330 | 79 | ||
| XM_021236859.1 | XP_021092518.1 | 9 - 10 | 8266 | 79 | ||
| XM_021236860.1 | XP_021092519.1 | 8 - 9 | 8126 | 79 | ||
| XM_021236861.1 | XP_021092520.1 | 7 - 8 | 8242 | 79 | ||

Tex15
Genomic Map