Assay Details
| Gene Symbol: Tex15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855457.2 | XP_004855514.2 | 8 - 9 | 9339 | 68 | |
| XM_021236851.1 | XP_021092510.1 | 10 - 11 | 9581 | 68 | ||
| XM_021236852.1 | XP_021092511.1 | 11 - 12 | 9703 | 68 | ||
| XM_021236853.1 | XP_021092512.1 | 11 - 12 | 9639 | 68 | ||
| XM_021236854.1 | XP_021092513.1 | 10 - 11 | 9499 | 68 | ||
| XM_021236855.1 | XP_021092514.1 | 9 - 10 | 9615 | 68 | ||
| XM_021236856.1 | XP_021092515.1 | 9 - 10 | 9505 | 68 | ||
| XM_021236857.1 | XP_021092516.1 | 9 - 10 | 9415 | 68 | ||
| XM_021236858.1 | XP_021092517.1 | 10 - 11 | 9537 | 68 | ||
| XM_021236859.1 | XP_021092518.1 | 10 - 11 | 9473 | 68 | ||
| XM_021236860.1 | XP_021092519.1 | 9 - 10 | 9333 | 68 | ||
| XM_021236861.1 | XP_021092520.1 | 8 - 9 | 9449 | 68 | ||
More Information
Set Membership: |
|

Tex15
Genomic Map