Assay Details
| Gene Symbol: Lrrn4cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874392.2 | XP_004874449.1 | 1 - 2 | 201 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004874392.2 transcripts.
Set Membership: |
|

Lrrn4cl
Genomic Map