Assay Details
| Gene Symbol: Znf527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239457.1 | XP_021095116.1 | 2 - 3 | 1044 | 91 | |
| XM_021239458.1 | XP_021095117.1 | 3 - 4 | 369 | 91 | ||
More Information
Set Membership: |
|

Znf527
Genomic Map