Assay Details
Gene Symbol: Cnot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004844451.3 | XP_004844508.1 | 4 - 5 | 1196 | 93 | |
XM_021258111.1 | XP_021113770.1 | 3 - 4 | 1055 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
