Assay Details
| Gene Symbol: Clic6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842585.3 | XP_004842642.1 | 5 - 6 | 1866 | 82 | |
| XM_021256210.1 | XP_021111869.1 | 5 - 6 | 697 | 82 | ||
| XM_021256211.1 | XP_021111870.1 | 5 - 6 | 609 | 82 | ||
More Information
Set Membership: |
|

Clic6
Genomic Map