Assay Details
Gene Symbol: Cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004841011.1 | XP_004841068.1 | 9 - 10 | 1276 | 65 | |
XM_004841013.3 | XP_004841070.1 | 9 - 10 | 1279 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
