Assay Details
| Gene Symbol: Ccl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843168.2 | XP_004843225.1 | 2 - 3 | 282 | 75 | |
| XM_021256485.1 | XP_021112144.1 | 1 - 2 | 552 | 75 | ||
More Information
Set Membership: |
|

Ccl17
Genomic Map