Assay Details
| Gene Symbol: Sntn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871611.3 | XP_004871668.1 | 3 - 4 | 594 | 68 | |
| XM_021249676.1 | XP_021105335.1 | 3 - 4 | 829 | 68 | ||
More Information
Set Membership: |
|

Sntn
Genomic Map